Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallikrein 5 Antibody (KLK5/3843), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB381125 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB381125 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB381125 Supplier Novus Biologicals Supplier No. NBP308365AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Kallikrein 5 Monoclonal antibody specifically detects Kallikrein 5 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen Kallikrein 5
Applications Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone KLK5/3843
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.21, EC 3.4.21.4, kallikrein 5, Kallikrein-like protein 2, kallikrein-related peptidase 5, KLKL2, KLK-L2EC 3.4.21.-, SCTEkallikrein-5, Stratum corneum tryptic enzyme
Host Species Mouse
Immunogen Recombinant fragment of human Kallikrein 5 protein (around aa 36-177) (exact sequence is proprietary) (Uniprot: Q9Y337)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 25818
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.