Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallikrein 5 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB344653 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344653 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344653 Supplier Novus Biologicals Supplier No. NB200138PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Kallikrein 5 Polyclonal antibody specifically detects Kallikrein 5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)

Specifications

Antigen Kallikrein 5
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation PBS
Gene Alias EC 3.4.21, EC 3.4.21.4, kallikrein 5, Kallikrein-like protein 2, kallikrein-related peptidase 5, KLKL2, KLK-L2EC 3.4.21.-, SCTEkallikrein-5, Stratum corneum tryptic enzyme
Host Species Rabbit
Immunogen Residues 280-293 [KFTKWIQETIQANS] of the human KLK-L2 protein.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 25818
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.