Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallikrein 6/Neurosin Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB052967 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB052967 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB052967 Supplier Novus Biologicals Supplier No. NBP298247AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Kallikrein 6/Neurosin Polyclonal specifically detects Kallikrein 6/Neurosin in Mouse samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Kallikrein 6/Neurosin
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias Bssp, EC 3.4.21, EC 3.4.21.-, hK6, kallikrein 6 (neurosin, zyme), kallikrein-6, kallikrein-related peptidase 6, Klk7, neurosin, Protease M, protease, serine, 18, PRSS18, PRSS9MGC9355, Serine protease 18, Serine protease 9, SP59, Zyme
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Kallikrein 6/Neurosin (NP_035307.2; Met 8-Leu 253).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5653
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.