Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallikrein 8/Neuropsin Antibody (140), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB036873 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036873 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036873 Supplier Novus Biologicals Supplier No. NBP289988IR
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Kallikrein 8/Neuropsin Monoclonal specifically detects Kallikrein 8/Neuropsin in Human samples. It is validated for Western Blot.

Specifications

Antigen Kallikrein 8/Neuropsin
Applications Western Blot
Classification Monoclonal
Clone 140
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.21, EC 3.4.21.118, hK8, HNP, kallikrein 8 (neuropsin/ovasin), kallikrein-related peptidase 8, KLK8, KLK8 protein type 1, KLK8 protein type 2, Neuropsin, neuropsin type 1, neuropsin type 2, NP, NRPN, Ovasin, PRSS19kallikrein-8, Serine protease 19, Serine protease TADG-14, TADG14neuropsin, Tumor-associated differentially expressed gene 14 protein
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Kallikrein 8/Neuropsin (O60259-1; Met1-Gly260).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 11202
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.