Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallikrein 8/Neuropsin Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB089938 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB089938 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB089938 Supplier Novus Biologicals Supplier No. NBP299878AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Kallikrein 8/Neuropsin Polyclonal specifically detects Kallikrein 8/Neuropsin in Human samples. It is validated for ELISA.

Specifications

Antigen Kallikrein 8/Neuropsin
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias EC 3.4.21, EC 3.4.21.118, hK8, HNP, kallikrein 8 (neuropsin/ovasin), kallikrein-related peptidase 8, KLK8, KLK8 protein type 1, KLK8 protein type 2, Neuropsin, neuropsin type 1, neuropsin type 2, NP, NRPN, Ovasin, PRSS19kallikrein-8, Serine protease 19, Serine protease TADG-14, TADG14neuropsin, Tumor-associated differentially expressed gene 14 protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Kallikrein 8/Neuropsin (O60259-1; Met 1-Gly 260).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 11202
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.