Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallikrein 9 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB018119 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018119 0.025 mg
NB200136 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018119 Supplier Novus Biologicals Supplier No. NB200136SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Kallikrein 9 Polyclonal specifically detects Kallikrein 9 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Kallikrein 9
Applications Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry, Immunohistochemistry (Frozen), ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500 -1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation PBS with 0.02% Sodium Azide
Gene Alias EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, kallikrein 9, Kallikrein-like protein 3, kallikrein-related peptidase 9, KLKL3, KLK-L3kallikrein-9, MGC50513
Gene Symbols KLK9
Host Species Rabbit
Immunogen Residues 239-250 [HYLDWIQEIMEN] of the human KLK-L3 (KLK9) protein
Molecular Weight of Antigen 28 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 284366
Test Specificity Reacts with residues 239-250 [HYLDWIQEIMEN] of the human KLK-L3 (KLK9) protein.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.