Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KANK2 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB185213 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB185213 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB185213 Supplier Novus Biologicals Supplier No. NBP282099AF350
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KANK2 Polyclonal antibody specifically detects KANK2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen KANK2
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ANKRD25, ankyrin repeat domain 25, Ankyrin repeat domain-containing protein 25, DKFZp434N161, FLJ20004, KIAA1518SRC1-interacting protein, kidney ankyrin repeat-containing protein 2, KN motif and ankyrin repeat domain-containing protein 2, KN motif and ankyrin repeat domains 2, Matrix-remodeling-associated protein 3, matrix-remodelling associated 3, MGC119707, MXRA3, SIP, SRC-1 interacting protein, SRC-1-interacting protein
Host Species Rabbit
Immunogen KANK2 antibody was raised against a 16 amino acid peptide near the carboxy terminus of human KANK2 . The immunogen is located within the last 50 amino acids of KANK2. Amino Acid Squence: CAPMSDDESPTSSSAEE
Purification Method Peptide Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 25959
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.