Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KANK3 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB185238 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB185238 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB185238 Supplier Novus Biologicals Supplier No. NBP282100AF594
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KANK3 Polyclonal antibody specifically detects KANK3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen KANK3
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ANKRD47, ankyrin repeat domain 47, Ankyrin repeat domain-containing protein 47, FLJ46061, kidney ankyrin repeat-containing protein 3, KN motif and ankyrin repeat domain-containing protein 3, KN motif and ankyrin repeat domains 3
Host Species Rabbit
Immunogen KANK3 antibody was raised against an 18 amino acid peptide near the amino terminus of human KANK3 . The immunogen is located within amino acids 50 - 100 of KANK3. Amino Acid Squence: CARRARRAPGPPTSRRPRA
Purification Method Peptide Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 256949
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.