Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KANK3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | KANK3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP156933
![]() |
Novus Biologicals
NBP156933 |
100 μL |
Each for $487.50
|
|
|||||
NBP15693320
![]() |
Novus Biologicals
NBP15693320UL |
20 μL | N/A | N/A | N/A | ||||
Description
KANK3 Polyclonal specifically detects KANK3 in Human samples. It is validated for Western Blot.Specifications
KANK3 | |
Polyclonal | |
Rabbit | |
Q6NY19-2 | |
256949 | |
Synthetic peptides corresponding to ANKRD47 The peptide sequence was selected from the N terminal of ANKRD47. Peptide sequence MAKFALNQNLPDLGGPRLCPVPAAGGARSPSSPYSVETPYGFHLDLDFLK. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
ANKRD47, ankyrin repeat domain 47, Ankyrin repeat domain-containing protein 47, FLJ46061, kidney ankyrin repeat-containing protein 3, KN motif and ankyrin repeat domain-containing protein 3, KN motif and ankyrin repeat domains 3 | |
KANK3 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title