Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KAO Monoclonal Antibody (C3), Invitrogen™
Mouse Monoclonal Antibody
$195.80 - $404.50
Specifications
Antigen | KAO |
---|---|
Clone | C3 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEMA551038
|
Invitrogen™
MA551038 |
20 μL |
Each for $195.80
|
|
|||||
FEMA551037
|
Invitrogen™
MA551037 |
200 μL |
Each for $404.50
|
|
|||||
Description
This gene encodes a metal-binding membrane glycoprotein that oxidatively deaminates putrescine, histamine, and related compounds. The encoded protein is inhibited by amiloride, a diuretic that acts by closing epithelial sodium ion channels. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Specifications
KAO | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
1600012D06Rik; Abp; abp1; amiloride binding protein 1 (amine oxidase (copper-containing)); amiloride binding protein 1 (amine oxidase, copper-containing); amiloride binding protein/diamine oxidase; amiloride-binding protein 1; amiloride-sensitive amine oxidase; amiloride-sensitive amine oxidase [copper-containing]; amine oxidase copper containing 1; Amine oxidase copper domain-containing protein 1; amine oxidase, copper containing 1; amine oxidase, copper-containing 1; aoc1; DAO; DAO1; Diamine oxidase; histaminase; KAO; kidney amine oxidase; si:ch211-286c5.2; zgc:154101 | |
AOC1 | |
IgG1 κ | |
Protein A/G | |
AOC1 |
C3 | |
Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
Unconjugated | |
Mouse | |
Human, Pig | |
P19801, Q9TRC7 | |
100517436, 26 | |
Recombinant Human ABP1 protein, Thr400-Val751 (Accession #P19801). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title