Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KAP1, p Ser824 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB290129 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290129 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290129 Supplier Novus Biologicals Supplier No. NBP232073AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KAP1 Polyclonal antibody specifically detects KAP1 in Mouse samples. It is validated for Western Blot, Immunoprecipitation (Negative)

Specifications

Antigen KAP1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunoprecipitation (Negative)
Formulation 50mM Sodium Borate
Gene Alias E3 SUMO-protein ligase TRIM28, EC 6.3.2.-, FLJ29029, KAP-1, KAP1KRAB-associated protein 1, KRIP-1, Nuclear corepressor KAP-1, RNF96KRAB-interacting protein 1, TF1B, TIF1-beta, TIF1BRING finger protein 96, transcription intermediary factor 1-beta, transcriptional intermediary factor 1-beta, tripartite motif containing 28, tripartite motif-containing 28, Tripartite motif-containing protein 28
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region of mouse KAP-1 (tripartite motif-containing 28 ) surrounding serine 824 using the numbering given in entry NP_035718.2 (Gene ID 21849) when the serine residue is phosphorylated.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Phospho Specific, Protein Kinase, Signal Transduction, Transcription Factors and Regulators, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 10155
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.