Learn More
Description
Specifications
Specifications
| Antigen | Kappa Light Chain |
| Applications | Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | KLC264 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunoprecipitation 1-2ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1ug/ml |
| Formulation | PBS with 0.05% BSA. with 0.05% Sodium Azide |
| Gene Accession No. | P01601 |
| Gene Alias | HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809 |
| Gene Symbols | IGKC |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
