Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kappa Light Chain Antibody (MEM-09), Biotin, Novus Biologicals™
SDP

Catalog No. NB359375 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359375 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359375 Supplier Novus Biologicals Supplier No. NB500331B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Kappa Light Chain Monoclonal antibody specifically detects Kappa Light Chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), CyTOF

Specifications

Antigen Kappa Light Chain
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone MEM-09
Conjugate Biotin
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, CyTOF-ready
Formulation PBS
Gene Alias HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809
Host Species Mouse
Immunogen Crude thymus membrane fraction.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3514
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.