Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Kartogenin
GREENER_CHOICE

Catalog No. p-4816671
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potently induces chondrogenesis in MSCs

Kartogenin potently induces differentiation of human mesenchymal stem cells into chondrocytes (EC50 = 100 nM). Reduces disease severity in a mouse model of osteoarthritis; displays protective effects against osteoarthritic stimuli in mature chondrocytes in vitro.

Chemical Identifiers

CAS 4727-31-5
Molecular Formula C20H15NO3

Specifications

Quantity 10 mg
Formula Weight 317.34
Purity >98%
Molecular Weight (g/mol) 317.344
Chemical Name or Material Kartogenin
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.