Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KAT2A/GCN5 Antibody (3G13), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP148593IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP148593IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP148593IR Supplier Novus Biologicals Supplier No. NBP148593IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KAT2A/GCN5 Monoclonal specifically detects KAT2A/GCN5 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen KAT2A/GCN5
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 3G13
Conjugate DyLight 755
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Gene Accession No. Q92830
Gene Alias EC 2.3.1.48, GCN5 general control of amino-acid synthesis 5-like 2 (yeast), GCN5L2GCN5 (general control of amino-acid synthesis, yeast, homolog)-like 2, GCN5MGC102791, General control of amino acid synthesis protein 5-like 2, General control of amino acid synthesis, yeast, homolog-like 2, HGCN5, Histone acetyltransferase GCN5, histone acetyltransferase KAT2A, HsGCN5, K(lysine) acetyltransferase 2A, Lysine acetyltransferase 2A, PCAF-b, STAF97
Gene Symbols KAT2A
Host Species Mouse
Immunogen Recombinant human GCN5L2 (411-837aa) purified from E. coli (NP_066564).
Molecular Weight of Antigen 94 kDa
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing, Epigenetics, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2648
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.