Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KAT3B/p300 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | KAT3B/p300 |
---|---|
Dilution | Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200, Immunocytochemistry |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
KAT3B/p300 Polyclonal specifically detects KAT3B/p300 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
KAT3B/p300 | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
E1A binding protein p300, E1A-associated protein p300, E1A-binding protein, 300kD, EC 2.3.1, EC 2.3.1.48, histone acetyltransferase p300, KAT3B, P300, p300 HAT, RSTS2 | |
EP300 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200, Immunocytochemistry | |
Polyclonal | |
Rabbit | |
Adaptive Immunity, Alzheimers Research, Amino Acids Drugs and other small molecules, Angiogenesis, Apoptosis, Asthma, Astrocyte Markers, AU1 and AU5 Epitope Tags, Autophagy, Base Excision Repair, Beta Galactosidase Epitope Tags, Breast Cancer, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, DNA Repair, HIF Target Genes, Hypoxia, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
2033 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:FLPQTQFPSQGMNVTNIPLAPSSGQAPVSQAQMSSSSCPVNSPIMPPGSQGSHIHCPQLPQPALHQNSPSPVPSRTPTPHHTPPSIGAQQPPATTIPAPVPTPPAMPPGPQSQALHPPPRQTPTPPTTQLPQQ | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title