Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KAT3B/p300 Antibody (RW109), Novus Biologicals™
SDP

Catalog No. NB100617 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100617 0.2 mL
NB100617SS 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100617 Supplier Novus Biologicals Supplier No. NB100617
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 4 publications

KAT3B/p300 Monoclonal specifically detects KAT3B/p300 in Human, Mouse, Mustelid, Primate, Rat samples. It is validated for Chemotaxis, ChIP Assay, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Western Blot.

Specifications

Antigen KAT3B/p300
Applications Chemotaxis, ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone RW109
Conjugate Unconjugated
Dilution Western Blot 1:1000, Chromatin Immunoprecipitation reported in scientific literature, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunoprecipitation 1:10-1:500, Chromatin Immunoprecipitation (ChIP)
Formulation Ascites with 0.1% Sodium Azide
Gene Alias E1A binding protein p300, E1A-associated protein p300, E1A-binding protein, 300kD, EC 2.3.1, EC 2.3.1.48, histone acetyltransferase p300, KAT3B, P300, p300 HAT, RSTS2
Gene Symbols EP300
Host Species Mouse
Immunogen Fusion protein containing residues 1572-2371 of human KAT3B/p300. [UniProt# Q09472]
Molecular Weight of Antigen 300 kDa
Purification Method Unpurified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Breast Cancer, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, DNA Repair, HIF Target Genes, Hypoxia, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 2033
Test Specificity This is specific for p300 protein. This recognizes residues 2107-2283.
Target Species Human, Mouse, Mouse, Mustelid, Primate, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.