Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KAT3B/p300 Antibody (RW109), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 4 publications
$206.50 - $499.50
Specifications
Antigen | KAT3B/p300 |
---|---|
Clone | RW109 |
Dilution | Western Blot 1:1000, Chromatin Immunoprecipitation reported in scientific literature, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunoprecipitation 1:10-1:500, Chromatin Immunoprecipitation (ChIP) |
Applications | Western Blot, ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100617SS
![]() |
Novus Biologicals
NB100617SS |
0.05 mL |
Each for $206.50
|
|
|||||
NB100617
![]() |
Novus Biologicals
NB100617 |
0.2 mL |
Each for $499.50
|
|
|||||
Description
KAT3B/p300 Monoclonal specifically detects KAT3B/p300 in Human, Mouse, Rat, Mustelid, Primate samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
KAT3B/p300 | |
Western Blot 1:1000, Chromatin Immunoprecipitation reported in scientific literature, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunoprecipitation 1:10-1:500, Chromatin Immunoprecipitation (ChIP) | |
Monoclonal | |
Ascites | |
RUO | |
Human, Mouse, Rat, Mustelid, Primate | |
E1A binding protein p300, E1A-associated protein p300, E1A-binding protein, 300kD, EC 2.3.1, EC 2.3.1.48, histone acetyltransferase p300, KAT3B, P300, p300 HAT, RSTS2 | |
EP300 | |
IgG1 κ | |
Unpurified | |
This is specific for p300 protein. This recognizes residues 2107-2283. |
RW109 | |
Western Blot, ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Apoptosis, Autophagy, Breast Cancer, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, DNA Repair, HIF Target Genes, Hypoxia, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors, Wnt Signaling Pathway | |
Ascites with 0.1% Sodium Azide | |
2033 | |
Fusion protein containing residues 1572-2371 of human KAT3B/p300. [UniProt# Q09472] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
300 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title