Learn More
Description
Specifications
Specifications
| Antigen | KAT3B/p300 |
| Applications | Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | RW128 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:250-1:500, Flow Cytometry, Immunocytochemistry/Immunofluorescence 2-5ug/mL, Immunoprecipitation 1:10-1:500, Flow (Intracellular) 1ug/mL |
| Gene Alias | E1A binding protein p300, E1A-associated protein p300, E1A-binding protein, 300kD, EC 2.3.1, EC 2.3.1.48, histone acetyltransferase p300, KAT3B, P300, p300 HAT, RSTS2 |
| Gene Symbols | EP300 |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
