Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KAT3B/p300 Antibody (RW128) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 7 publications

$207.00 - $499.50

Specifications

Antigen KAT3B/p300
Clone RW128
Concentration 1 mg/mL
Dilution Western Blot 1:250-1:500, Flow Cytometry, Immunocytochemistry/Immunofluorescence 2-5ug/mL, Immunoprecipitation 1:10-1:500, Flow (Intracellular) 1ug/mL
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB100507SS
SDP
View Documents
Novus Biologicals
NB100507SS
0.025 mg
Each for $207.00
Only null left
Add to Cart
 
NB100507
SDP
View Documents
Novus Biologicals
NB100507
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

KAT3B/p300 Monoclonal specifically detects KAT3B/p300 in Human, Mouse, Rat, Mustelid, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Flow (Intracellular).
Specifications

Specifications

KAT3B/p300
1 mg/mL
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
Mouse
Apoptosis, Autophagy, Breast Cancer, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, DNA Repair, HIF Target Genes, Hypoxia, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors, Wnt Signaling Pathway
E1A binding protein p300, E1A-associated protein p300, E1A-binding protein, 300kD, EC 2.3.1, EC 2.3.1.48, histone acetyltransferase p300, KAT3B, P300, p300 HAT, RSTS2
EP300
IgG1 κ
Protein A purified
300 kDa
RW128
Western Blot 1:250-1:500, Flow Cytometry, Immunocytochemistry/Immunofluorescence 2-5ug/mL, Immunoprecipitation 1:10-1:500, Flow (Intracellular) 1ug/mL
Monoclonal
Purified
RUO
Human, Mouse, Rat, Mustelid, Primate
2033
Fusion protein containing residues 1572-2371 of human KAT3B/p300. [UniProt# Q0947]
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.