Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KAT3B/p300 Mouse anti-Human, Mouse, Rat, Mustelid, Primate, DyLight 550, Clone: RW128, Novus Biologicals™
SDP

Catalog No. NB170791 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170791 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB170791 Supplier Novus Biologicals Supplier No. NB100507R0.1ML
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

KAT3B/p300 Monoclonal antibody specifically detects KAT3B/p300 in Human, Mouse, Rat, Mustelid, Primate samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen KAT3B/p300
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone RW128
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias E1A binding protein p300, E1A-associated protein p300, E1A-binding protein, 300kD, EC 2.3.1, EC 2.3.1.48, histone acetyltransferase p300, KAT3B, P300, p300 HAT, RSTS2
Host Species Mouse
Immunogen Fusion protein containing residues 1572-2371 of human KAT3B/p300. [UniProt# Q0947]
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Breast Cancer, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, DNA Repair, HIF Target Genes, Hypoxia, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 2033
Target Species Human, Mouse, Rat, Mustelid, Primate
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.