Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KAT4/TBP Associated Factor 1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP177847 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177847 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177847 Supplier Novus Biologicals Supplier No. NBP177847
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KAT4/TBP Associated Factor 1 Polyclonal specifically detects KAT4/TBP Associated Factor 1 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen KAT4/TBP Associated Factor 1
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:150000-1:300000, Immunohistochemistry 10ug/ml, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 10 ug/ml, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. P21675
Gene Alias 250kDa, BA2Rdystonia 3 (with Parkinsonism), CCG1TATA box binding protein (TBP)-associated factor, RNA polymerase II, A, 250kD, CCGS, Cell cycle gene 1 protein, cell cycle, G1 phase defect, complementation of cell cycle block, G1-to-S, DYT3/TAF1, EC 2.7.11, EC 2.7.11.1, KAT4, NSCL2, OF, P250, TAF(II)250, TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF2AN-TAF1, TAFII-250, TAFII250DYT3, TBP-associated factor 250 kDa, transcription factor TFIID p250 polypeptide, Transcription initiation factor TFIID 250 kDa subunit, transcription initiation factor TFIID subunit 1, XDP
Gene Symbols TAF1
Host Species Rabbit
Immunogen This affinity purified KAT4/TBP Associated Factor 1 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids near the carboxyl terminus of human KAT4/TBP Associated Factor 1. (Uniprot: P21675)
Molecular Weight of Antigen 212.68 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 6872
Test Specificity A BLAST analysis was used to suggest cross-reactivity with TAF1 protein from human, mouse and rat based on 100% homology with the immunizing sequence. Reactivity against homologues from other sources is not known.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.