Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KAT4/TBP Associated Factor 1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB093635 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB093635 20 μL
NB093634 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB093635 Supplier Novus Biologicals Supplier No. NBP30349920UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KAT4/TBP Associated Factor 1 Polyclonal specifically detects KAT4/TBP Associated Factor 1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen KAT4/TBP Associated Factor 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias 250kDa, BA2Rdystonia 3 (with Parkinsonism), CCG1TATA box binding protein (TBP)-associated factor, RNA polymerase II, A, 250kD, CCGS, Cell cycle gene 1 protein, cell cycle, G1 phase defect, complementation of cell cycle block, G1-to-S, DYT3/TAF1, EC 2.7.11, EC 2.7.11.1, KAT4, NSCL2, OF, P250, TAF(II)250, TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF2AN-TAF1, TAFII-250, TAFII250DYT3, TBP-associated factor 250 kDa, transcription factor TFIID p250 polypeptide, Transcription initiation factor TFIID 250 kDa subunit, transcription initiation factor TFIID subunit 1, XDP
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1630-1893 of human KAT4/TBP Associated Factor 1 (NP_004597.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 6872
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.