Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KAT6B-MORF Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB092889 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB092889 20 μL
NB092888 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB092889 Supplier Novus Biologicals Supplier No. NBP30299020UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KAT6B-MORF Polyclonal specifically detects KAT6B-MORF in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen KAT6B-MORF
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias DKFZp313G1618, FLJ90335, Histone acetyltransferase MORF, Histone acetyltransferase MOZ2, histone acetyltransferase MYST4, KAT6B, KIAA0383, Monocytic leukemia zinc finger protein-related factor, MORFMorf, MOZ, YBF2/SAS3, SAS2 and TIP60 protein 4, MOZ2EC 2.3.1.48, MOZ-related factor, MYST histone acetyltransferase (monocytic leukemia) 4, MYST-4, qkf, querkopf
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 400-500 of human KAT6B-MORF (NP_001243398.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23522
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.