Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Katanin p60 Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB329782 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB329782 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB329782 Supplier Novus Biologicals Supplier No. NBP297830JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Katanin p60 Polyclonal antibody specifically detects Katanin p60 in Human samples. It is validated for Western Blot

Specifications

Antigen Katanin p60
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias EC 3.6.4.3, katanin p60 (ATPase containing) subunit A 1, katanin p60 (ATPase-containing) subunit A 1, katanin p60 ATPase-containing subunit A1, Katanin p60 subunit A1, p60 katanin
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Katanin p60 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 11104
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.