Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

katanin-p80 Antibody (OTI5C4), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB271578A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB271578A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB271578A48 Supplier Novus Biologicals Supplier No. NBP271578AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

katanin-p80 Monoclonal specifically detects katanin-p80 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen katanin-p80
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI5C4
Conjugate Alexa Fluor 488
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias KAT, katanin (80 kDa), katanin p80 (WD repeat containing) subunit B 1, katanin p80 (WD40-containing) subunit B 1, Katanin p80 subunit B1, katanin p80 WD40-containing subunit B1, p80 katanin
Gene Symbols KATNB1
Host Species Mouse
Immunogen Full length recombinant protein of human katanin-p80 (NP_005877) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10300.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.