Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KATNA1 Polyclonal Antibody
Rabbit Polyclonal Antibody
$237.00 - $511.50
Specifications
Antigen | KATNA1 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119239
|
Invitrogen™
PA5119239 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121810
|
Invitrogen™
PA5121810 |
20 μL |
Each for $237.00
|
|
|||||
Description
KATNA1 Polyclonal Antibody for Western Blot, IHC (P)
Microtubules, polymers of alpha and beta tubulin subunits, form the mitotic spindle of a dividing cell and help to organize membranous organelles during interphase. Katanin is a heterodimer that consists of a 60 kDa ATPase (p60 subunit A 1) and an 80 kDa accessory protein (p80 subunit B 1). The p60 subunit acts to sever and disassemble microtubules, while the p80 subunit targets the enzyme to the centrosome. This gene encodes the p80 subunit. This protein is a member of the AAA family of ATPases.Specifications
KATNA1 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human, Mouse | |
KATNA1 | |
HAMAP-Rule:MF_03023}; hypothetical protein LOC553631; katanin catalytic subunit A1; katanin p60 (ATPase containing) subunit A 1; katanin p60 (ATPase-containing) subunit A 1; katanin p60 (ATPase-containing) subunit A1; katanin p60 ATPase-containing subunit A1; katanin p60 ATPase-containing subunit A1 {ECO:0000255; Katanin p60 ATPase-containing subunit A1-like protein; katanin p60 subunit A 1; katanin p60 subunit A1; katanin p60 subunit A1 {ECO:0000255; KATNA1; lipotransin; microtubule severing protein; p60 katanin; p60 katanin {ECO:0000255; si:dkey-15j16.1; zgc:110580 | |
KATNA1 | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
O75449, Q9WV86 | |
11104, 23924 | |
Recombinant protein Katanin P60 Subunit A 1. The antigen corresponds to amino acid range 260-491 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title