Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KATNAL1 Antibody (OTI1A7), PerCP, Novus Biologicals™
SDP

Catalog No. NB272254PCP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB272254PCP 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB272254PCP Supplier Novus Biologicals Supplier No. NBP272254PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KATNAL1 Monoclonal specifically detects KATNAL1 in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Flow Cytometry.

Specifications

Antigen KATNAL1
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1A7
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 3.6.4.3, katanin p60 ATPase-containing subunit A-like 1, katanin p60 subunit A-like 1MGC2599, p60 katanin-like 1
Gene Symbols KATNAL1
Host Species Mouse
Immunogen Full length recombinant protein of human KATNAL1 (NP_115492) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 84056.0
Target Species Human, Mouse, Rat, Primate, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.