Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KAZALD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
$237.00 - $539.50
Specifications
Antigen | KAZALD1 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119241
|
Invitrogen™
PA5119241 |
200 μL |
Each for $539.50
|
|
|||||
PIPA5121808
|
Invitrogen™
PA5121808 |
20 μL |
Each for $237.00
|
|
|||||
Description
KAZALD1 Polyclonal Antibody for Western Blot, IHC (P)
This gene encodes a secreted member of the insulin growth factor-binding protein (IGFBP) superfamily. It contains an N-terminal insulin growth factor-binding domain, a central Kazal-type serine protease inhibitor and follistatin-like domain, and a C-terminal immunoglobulin-like domain. Studies of the mouse ortholog suggest that this gene product may have a function in bone development and bone regeneration.Specifications
KAZALD1 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human, Bovine, Pig | |
KAZALD1 | |
bone- and odontoblast-expressed gene 1; BONO1; FKSG28; FKSG40; FLJ24094; IGFBP-related protein 10; IGFBP-rP10; Kazal type serine peptidase inhibitor domain 1; KAZALD1; kazal-type serine protease inhibitor domain-containing protein 1; novel kazal-type serine protease inhibitor domain and immunoglobulin domain containing protein; UNQ2945/PRO21184 | |
KAZALD1 | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q96I82 | |
100624902, 617445, 81621 | |
Recombinant protein Kazal Type Serine Protease Inhibitor Domain Containing Protein 1. The antigen corresponds to amino acid range 31-304 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title