Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KBTBD7 Antibody (OTI1G3), PerCP, Novus Biologicals™
SDP

Catalog No. NB288281 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288281 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288281 Supplier Novus Biologicals Supplier No. NBP272268PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KBTBD7 Monoclonal antibody specifically detects KBTBD7 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen KBTBD7
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI1G3
Conjugate PerCP
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias DKFZP434E2318, kelch repeat and BTB (POZ) domain containing 7, kelch repeat and BTB domain-containing protein 7
Host Species Mouse
Immunogen Full length human recombinant protein of human KBTBD7 (NP_115514) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 84078
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.