Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ KC7F2
GREENER_CHOICE

Catalog No. 432410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-241-0 10 mg
43-245-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 43-241-0 Supplier Tocris Bioscience™ Supplier No. 4324/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

HIF-1α inhibitor; down-regulates HIF-1α protein synthesis

Inhibitor of HIF-1α. Thought to act via down-regulation of HIF-1α protein synthesis; reduces phosphorylation of eIF4E binding protein 1 (4EBP1) and p70 S6K in hypoxic conditions. Also blocks hypoxia-induced HIF-1α accumulation in a range of human cancer cell lines. Inhibits the expression of HIF target genes, such as carbonic anhydrase IX and MMP2.

Chemical Identifiers

CAS 927822-86-4
Molecular Formula C16H16Cl4N2O4S4

Specifications

Quantity 10 mg
Formula Weight 570.38
Purity >98%
Molecular Weight (g/mol) 570.354
Chemical Name or Material KC7F2
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.