Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCC1/SLC12A4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | KCC1/SLC12A4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
KCC1/SLC12A4 Polyclonal specifically detects KCC1/SLC12A4 in Human samples. It is validated for Western Blot.Specifications
KCC1/SLC12A4 | |
Polyclonal | |
Rabbit | |
Q9UP95 | |
6560 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
Electroneutral potassium-chloride cotransporter 1, Erythroid K-Cl cotransporter 1, FLJ17069, FLJ40489, hKCC1, KCC1erythroid K:Cl cotransporter, K-Cl cotransporter, potassium/chloride cotransporter 1, solute carrier family 12 (potassium/chloride transporters), member 4, solute carrier family 12 member 4 | |
SLC12A4 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title