Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCC2/SLC12A5 Antibody (905705), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB372994 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB372994 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB372994 Supplier Novus Biologicals Supplier No. FAB8369Z
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KCC2/SLC12A5 Monoclonal specifically detects KCC2/SLC12A5 in Human samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen KCC2/SLC12A5
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 905705
Conjugate DyLight 755
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Electroneutral potassium-chloride cotransporter 2, hKCC2, KCC2K-Cl cotransporter 2, KIAA1176erythroid K-Cl cotransporter 2, Neuronal K-Cl cotransporter, solute carrier family 12 (potassium/chloride transporter), member 5, solute carrier family 12 member 5
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human SLC12A5, Accession # Q9H2X9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Potassium Channels
Primary or Secondary Primary
Gene ID (Entrez) 57468
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.