Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCC2/SLC12A5 Antibody (S1-12), Biotin, Novus Biologicals™
SDP

Catalog No. NBP259337B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259337B 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259337B Supplier Novus Biologicals Supplier No. NBP259337B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KCC2/SLC12A5 Monoclonal specifically detects KCC2/SLC12A5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen KCC2/SLC12A5
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S1-12
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias Electroneutral potassium-chloride cotransporter 2, hKCC2, KCC2K-Cl cotransporter 2, KIAA1176erythroid K-Cl cotransporter 2, Neuronal K-Cl cotransporter, solute carrier family 12 (potassium/chloride transporter), member 5, solute carrier family 12 member 5
Gene Symbols SLC12A5
Host Species Mouse
Immunogen Fusion protein amino acids 932-1043 corresponding to rat KCC2
Purification Method Protein G purified
Quantity 100 μL
Research Discipline Potassium Channels
Primary or Secondary Primary
Gene ID (Entrez) 57468
Test Specificity Detects 140kDa.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.