Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCC2/SLC12A5, p Ser940 Antibody, Novus Biologicals™
SDP

Catalog No. NBP229513 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229513 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP229513 Supplier Novus Biologicals Supplier No. NBP229513
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

KCC2/SLC12A5 Polyclonal specifically detects KCC2/SLC12A5 in Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen KCC2/SLC12A5 (p Ser940)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative
Gene Accession No. Q63633
Gene Alias Electroneutral potassium-chloride cotransporter 2, hKCC2, KCC2K-Cl cotransporter 2, KIAA1176erythroid K-Cl cotransporter 2, Neuronal K-Cl cotransporter, solute carrier family 12 (potassium/chloride transporter), member 5, solute carrier family 12 member 5
Gene Symbols SLC12A5
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser940 conjugated to KLH. Accession # Q63633
Molecular Weight of Antigen 135 kDa
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Potassium Channels
Primary or Secondary Primary
Gene ID (Entrez) 57468
Test Specificity Specific for the ∼135k KCC2 protein phosphorylated at Ser940. Immunolabeling of the KCC2 protein band is blocked by the phospho-peptide used as antigen but not by the corresponding dephosphopeptide.
Target Species Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.