Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCC4/SLC12A7 Antibody (891526), HRP, Novus Biologicals™
SDP

Catalog No. NB345895 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345895 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345895 Supplier Novus Biologicals Supplier No. FAB9030H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KCC4/SLC12A7 Monoclonal specifically detects KCC4/SLC12A7 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen KCC4/SLC12A7
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 891526
Conjugate HRP
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias DKFZp434F076, KCC4Electroneutral potassium-chloride cotransporter 4, K-Cl cotransporter 4, potassium/chloride transporter KCC4, solute carrier family 12 (potassium/chloride transporters), member 7, solute carrier family 12 member 7
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human SLC12A7, Met1-Ser1083, Accession # Q9Y666
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10723
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.