Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KChIP2 Antibody (OTI3C4), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP271866FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271866FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271866FR Supplier Novus Biologicals Supplier No. NBP271866FR

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

KChIP2 Monoclonal specifically detects KChIP2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen KChIP2
Applications Western Blot
Classification Monoclonal
Clone OTI3C4
Conjugate DyLight 680
Dilution Western Blot
Formulation PBS with 0.05% Sodium Azide
Gene Alias A-type potassium channel modulatory protein 2, cardiac voltage gated potassium channel modulatory subunit, Cardiac voltage-gated potassium channel modulatory subunit, DKFZp566L1246, KChIP2, KCHIP2Kv channel-interacting protein 2, Kv channel interacting protein 2, MGC17241, potassium channel interacting protein 2, Potassium channel-interacting protein 2
Gene Symbols KCNIP2
Host Species Mouse
Immunogen Full length recombinant protein of human KChIP2 (NP_775284) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 30819.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.