Learn More
Description
Specifications
Specifications
| Antigen | KCNA10 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | cyclic GMP gated potassium channel, Kcn1, Kv1.8, potassium voltage-gated channel subfamily A member 10, potassium voltage-gated channel, shaker-related subfamily, member 10, Voltage-gated potassium channel subunit Kv1.8 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the N terminal region of human KCNA10 (NP_005540). Peptide sequence DVCGWKEMEVALVNFDNSDEIQEEPGYATDFDSTSPKGRPGGSSFSNGKI |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
