Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNA7 Antibody, Novus Biologicals™
SDP

Catalog No. NBP118993 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP118993 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP118993 Supplier Novus Biologicals Supplier No. NBP118993
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KCNA7 Polyclonal specifically detects KCNA7 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen KCNA7
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:200-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1-:50 - 1:200, Immunohistochemistry-Paraffin 1:50-1:200
Formulation 0.1M Tris 0.1M Glycine and 2% Sucrose with No Preservative
Gene Accession No. Q96RP8
Gene Alias HAK6, KV1.7, potassium voltage-gated channel subfamily A member 7, potassium voltage-gated channel, shaker-related subfamily, member 7, voltage-dependent potassium channel Kv1.7, voltage-gated potassium channel KCNA7, Voltage-gated potassium channel subunit Kv1.7
Gene Symbols KCNA7
Host Species Rabbit
Immunogen Synthetic peptide derived from C-terminal domain of KCNA7 protein
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3743
Test Specificity Reacts specifically with human 50 kDa protein
Reconstitution Reconstitute with distilled water.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.