Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB151753 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151753 100 μg
NB151754 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB151753 Supplier Novus Biologicals Supplier No. NBP311900100UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KCNB1 Polyclonal antibody specifically detects KCNB1 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen KCNB1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 5 - 10 ug/ml
Formulation PBS
Gene Alias Delayed rectifier potassium channel 1, delayed rectifier potassium channel Kv2.1, DRK1, h-DRK1, h-DRK1 K(+) channel, Kv2.1, potassium channel protein DRK1, potassium voltage-gated channel subfamily B member 1, potassium voltage-gated channel, Shab-related subfamily, member 1, Voltage-gated potassium channel subunit Kv2.1
Host Species Rabbit
Immunogen Rabbits were immunized with a synthetic peptide derived from the rat Kv2.1 potassium channel conjugated to KLH (Uniprot: Q14721)
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3745
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.