Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCNH1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | KCNH1 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
KCNH1 Polyclonal specifically detects KCNH1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
KCNH1 | |
Polyclonal | |
Rabbit | |
Human | |
3756 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:DSCDSGITKSDLRLDNVGEARSPQDRSPILAEVKHSFYPIPEQTLQATVLEVRHELKEDIKALNAKMTNIEKQLSEILRILTSRRSSQSPQELFEISRPQSPES | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
EAG channel 1, EAG1, EAGeag1, Ether-a-go-go potassium channel 1, ether-a-go-go, Drosophila, homolog of, hEAG1, h-eageag, Kv10.1, MGC142269, potassium channel, voltage-gated, subfamily H, member 1, potassium voltage-gated channel subfamily H member 1, potassium voltage-gated channel, subfamily H (eag-related), member 1, Voltage-gated potassium channel subunit Kv10.1 | |
KCNH1 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title