Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNJ10 Antibody, Novus Biologicals™
SDP

Catalog No. NB018397 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018397 0.025 mL
NBP120149 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018397 Supplier Novus Biologicals Supplier No. NBP1201490.025ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KCNJ10 Polyclonal specifically detects KCNJ10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen KCNJ10
Applications Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry (Frozen), Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:300-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation Whole antisera with No Preservative
Gene Accession No. P78508
Gene Alias ATP-dependent inwardly rectifying potassium channel Kir4.1, ATP-sensitive inward rectifier potassium channel 10, BIRK-10, glial ATP-dependent inwardly rectifying potassium channel KIR4.1, Inward rectifier K(+) channel Kir1.2, inward rectifier K+ channel KIR1.2, KCNJ13-PEN, KIR1.2, Kir4.1, Potassium channel, inwardly rectifying subfamily J member 10, potassium inwardly-rectifying channel, subfamily J, member 10, SESAME
Gene Symbols KCNJ10
Host Species Rabbit
Immunogen A synthetic peptide from the c-terminal region of human KCNJ10 (Kir1.2) conjugated to an immunogenic carrier protein was used as the immunogen. The antigen is homologous in mouse and rat.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3766
Test Specificity KCNJ10.
Reconstitution Reconstitute with 0.1 ml sterilized water to desired concentration.
Target Species Rat, Mouse, Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.