Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNJ13 Rabbit anti-Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB096090 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB096090 20 μL
NB096089 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB096090 Supplier Novus Biologicals Supplier No. NBP30475120UL

Rabbit Polyclonal Antibody

KCNJ13 Polyclonal specifically detects KCNJ13 in Rat samples. It is validated for Western Blot.

Specifications

Antigen KCNJ13
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias Inward rectifier K(+) channel Kir7.1, inwardly rectifying subfamily J member 13, MGC33328, potassium inwardly-rectifying channel, subfamily J, member 13
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 160-360 of human KCNJ13 (NP_002233.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 3769
Target Species Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.