Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNK1 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB202194 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB202194 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB202194 Supplier Novus Biologicals Supplier No. NBP241301H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KCNK1 Polyclonal antibody specifically detects KCNK1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen KCNK1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias DPK, HOHO1, Inward rectifying potassium channel protein TWIK-1, K2P1, K2p1.1, KCNO1, Potassium channel KCNO1, potassium channel subfamily K member 1, potassium channel, subfamily K, member 1, potassium inwardly-rectifying channel, subfamily K, member 1, TWIK-1, TWIK1HOHO
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid peptide near the carboxy terminus of human KCNK1. The immunogen is located within amino acids 220 - 270 of KCNK1. Amino Acid Squence: DYVPGEGYNQKFRELYK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3775
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.