Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNK12 Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB164154 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164154 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB164154 Supplier Novus Biologicals Supplier No. NBP281727AF405
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KCNK12 Polyclonal antibody specifically detects KCNK12 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen KCNK12
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias potassium channel subfamily K member 12, potassium channel, subfamily K, member 12, tandem pore domain potassium channel THIK-2, THIK2, THIK-2, THIK-2THIK2Tandem pore domain halothane-inhibited potassium channel 2
Host Species Rabbit
Immunogen KCNK12 antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human KCNK12. The immunogen is located within amino acids 320 - 370 of KCNK12. Amino Acid Squence: PGSRLRRRLAALGA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 56660
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.