Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNK12 Antibody, Novus Biologicals™
SDP

Catalog No. NBP18008420 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP18008420 20 μL
NBP180084 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP18008420 Supplier Novus Biologicals Supplier No. NBP18008420UL

Rabbit Polyclonal Antibody

KCNK12 Polyclonal specifically detects KCNK12 in Human samples. It is validated for Western Blot.

Specifications

Antigen KCNK12
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. NP_071338
Gene Alias potassium channel subfamily K member 12, potassium channel, subfamily K, member 12, tandem pore domain potassium channel THIK-2, THIK2, THIK-2, THIK-2THIK2Tandem pore domain halothane-inhibited potassium channel 2
Gene Symbols KCNK12
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human KCNK12. Peptide sequence EGRRLSGELISMRDLTASNKVSLALLQKQLSETANGYPRSVCVNTRQNGF.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56660
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.