Learn More
Description
Specifications
Specifications
| Antigen | KCNK3 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | S374-48 |
| Conjugate | DyLight 550 |
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | Acid-sensitive potassium channel protein TASK-1, cardiac potassium channel, K2p3.1, OAT1, potassium channel subfamily K member 3, potassium channel, subfamily K, member 3, potassium inwardly-rectifying channel, subfamily K, member 3, TASK1, TASK-1, TASKacid-sensitive potassium channel protein TASK, TBAK1, TWIK-related acid-sensitive K(+) channel 1, TWIK-related acid-sensitive K+ channel, two P domain potassium channel, Two pore K(+) channel KT3.1, Two pore potassium channel KT3.1 |
| Gene Symbols | KCNK3 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
