Learn More
Description
Specifications
Specifications
| Antigen | KCNK4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose |
| Gene Alias | K2p4.1, K2P4.1 potassium channel, potassium channel subfamily K member 4, potassium channel, subfamily K, member 4, TRAAKTRAAK1, TWIK-related arachidonic acid-stimulated potassium channel protein, Two pore K(+) channel KT4.1, two pore K+ channel KT4.1, Two pore potassium channel KT4.1 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of human KCNK4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
