Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNK9 Antibody, Novus Biologicals™
SDP

Catalog No. NB018621 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018621 0.025 mL
NBP146493 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018621 Supplier Novus Biologicals Supplier No. NBP1464930.025ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KCNK9 Polyclonal specifically detects KCNK9 in Mouse samples. It is validated for Immunohistochemistry.

Specifications

Antigen KCNK9
Applications Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 1:10-1:500
Formulation Whole antisera with No Preservative
Gene Alias KT3.2, potassium channel, subfamily K, member 9, TASK-3
Gene Symbols KCNK9
Host Species Rabbit
Immunogen A synthetic peptide from the c-terminal region of mouse KCNK9 (TASK3) conjugated to an immunogenic carrier protein was used as the immunogen.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51305
Test Specificity Specific for KCNK9.
Reconstitution Reconstitute with 0.1 ml sterilized water to desired concentration.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.