Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNK9 Antibody, Novus Biologicals™
SDP

Catalog No. NBP146493 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP146493 0.1 mL
NB018621 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP146493 Supplier Novus Biologicals Supplier No. NBP146493
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KCNK9 Polyclonal specifically detects KCNK9 in Mouse samples. It is validated for Immunohistochemistry.

Specifications

Antigen KCNK9
Applications Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 1:300-1:2000
Formulation Lyophilized from whole antisera with No Preservative
Gene Alias KT3.2, potassium channel, subfamily K, member 9, TASK-3
Gene Symbols KCNK9
Host Species Rabbit
Immunogen A synthetic peptide from mouse KCNK9 conjugated to blue carrier protein was used as the antigen.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51305
Test Specificity Specific for KCNK9.
Reconstitution Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.